N-Boc-5-bromo-2,3-difluoroaniline 95%

Supplier: Ambeed
Warning

A549654-100MG A549654-250MG
76878-474EA 100 USD
76878-474 76878-476
N-Boc-5-bromo-2,3-difluoroaniline 95%
N-Boc-5-bromo-2,3-difluoroaniline

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrF₂NO₂
MDL Number: MFCD12025956
CAS Number: 1150114-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR