2-(Trifluoromethoxy)ethylamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Trifluoromethoxy)ethanamine hydrochloride

A548560-100MG A548560-1G A548560-250MG A548560-5G
76878-466EA 16 USD
76878-466 76878-468 76878-470 76878-472
2-(Trifluoromethoxy)ethylamine hydrochloride 95%
2-(Trifluoromethoxy)ethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₆F₃NO·ClH
MW: 165.542 g/mol
MDL Number: MFCD08447280
CAS Number: 886050-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR