2-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
Supplier: Ambeed
|
Warning
|
A547990-100MG
A547990-1G
A547990-250MG
A547990-5G
76878-458EA
11
USD
76878-458
76878-460
76878-462
76878-464
2-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₃H₁₄BF₅O₂ |
MDL Number:
MFCD18434440 CAS Number: 881402-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...