3,5-Difluoro-2-nitropyridine 95%

Supplier: Ambeed
Danger

A545394-1G A545394-250MG
76878-446EA 597.29 USD
76878-446 77613-722
3,5-Difluoro-2-nitropyridine 95%
3,5-Difluoro-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂F₂N₂O₂
MW: 160.07 g/mol
MDL Number: MFCD18802625
CAS Number: 1073634-13-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR