4-Bromo-N-cyclopropyl-3-methylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

A545230-1G A545230-250MG
76878-442EA 44.01 USD
76878-442 76878-444
4-Bromo-N-cyclopropyl-3-methylbenzenesulfonamide 98%
4-Bromo-N-cyclopropyl-3-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂S
MDL Number: MFCD07838964
CAS Number: 876875-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR