1-(2,4-Dichloropyridin-3-yl)ethanone 97%

Supplier: Ambeed
Warning

A544845-250MG A544845-5G A544845-1G
76878-434EA 130.5 USD
76878-434 76878-436 77613-716
1-(2,4-Dichloropyridin-3-yl)ethanone 97%
1-(2,4-Dichloropyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO
MW: 190.02 g/mol
MDL Number: MFCD11847304
CAS Number: 1246349-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR