1-(6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazol-1-yl)ethanone 98%

Supplier: Ambeed
Danger

A543731-1G A543731-250MG A543731-5G
76878-424EA 160.2 USD
76878-424 76878-426 76878-428
1-(6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazol-1-yl)ethanone 98%
1-(6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazol-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₉BN₂O₃
MW: 286.13 g/mol
MDL Number: MFCD12032487
CAS Number: 1256359-07-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR