tert-Butyl 2-chloro-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 96%

Supplier: Ambeed
Warning

A542777-1G A542777-250MG A542777-5G
76878-410EA 208 USD
76878-410 76878-412 76878-414
tert-Butyl 2-chloro-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 96%
tert-Butyl 2-chloro-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₇H₂₃BClFO₄
MW: 356.63 g/mol
MDL Number: MFCD15143607
CAS Number: 1218790-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR