4'-Amino-3'-cyanoacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 5-Acetyl-2-aminobenzonitrile

A541839-1G A541839-250MG A541839-5G
76878-400EA 82.76 USD
76878-400 76878-402 76878-404
4'-Amino-3'-cyanoacetophenone 98%
4'-Amino-3'-cyanoacetophenone

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.1726 g/mol
MDL Number: MFCD18379155
CAS Number: 33720-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR