5-Nitro-1H-imidazole-4-carbaldehyde 95%

Supplier: Ambeed
Warning

A541431-250MG A541431-1G
76878-398EA 140 USD
76878-398 77569-056
5-Nitro-1H-imidazole-4-carbaldehyde 95%
5-Nitro-1H-imidazole-4-carbaldehyde

Precaución: For research use only.

Formula: C₄H₃N₃O₃
MDL Number: MFCD09743898
CAS Number: 81246-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR