4-Bromo-4'-fluorobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)(4-fluorophenyl)methanone

A538536-1G A538536-250MG A538536-5G
76878-388EA 45 USD
76878-388 76878-390 76878-392
4-Bromo-4'-fluorobenzophenone 98%
4-Bromo-4'-fluorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₈BrFO
MDL Number: MFCD00672035
CAS Number: 2069-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR