4-(2-Fluorophenyl)-3,4-dihydronaphthalen-1(2H)-one 95%

Supplier: Ambeed
Warning

A537865-1G A537865-5G
76878-384EA 64.72 USD
76878-384 76878-386
4-(2-Fluorophenyl)-3,4-dihydronaphthalen-1(2H)-one 95%
4-(2-Fluorophenyl)-3,4-dihydronaphthalen-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₆H₁₃FO
MW: 240.27 g/mol
MDL Number: MFCD22421610
CAS Number: 82101-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR