2-Methylcyclobutanecarboxylic acid 98%

Supplier: Ambeed
Warning

A536446-100MG A536446-1G A536446-250MG
76878-382EA 86 USD
76878-382 77673-434 77884-332
2-Methylcyclobutanecarboxylic acid 98%
2-Methylcyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MDL Number: MFCD09266201
CAS Number: 42185-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR