3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-3-methylpyridine-5-boronic acid, pinacol ester

A535488-100MG A535488-1G
76878-368EA 26 USD
76878-368 76878-370
3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile 95%
3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₂O₂
MW: 244.09 g/mol
MDL Number: MFCD12026105
CAS Number: 1150561-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR