4,5-Difluorooxindole 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluoroindolin-2-one

A535103-100MG A535103-1G A535103-250MG
76878-356EA 37.14 USD
76878-356 76878-358 76878-360
4,5-Difluorooxindole 97%
4,5-Difluorooxindole

Precaución: for Research Use Only

Formula: C₈H₅F₂NO
MW: 169.13 g/mol
MDL Number: MFCD04112481
CAS Number: 850429-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR