7-Aminoquinazoline-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A532977-100MG A532977-1G A532977-250MG A532977-25G A532977-5G
76878-336EA 15.92 USD
76878-336 76878-338 76878-340 76878-342 76878-344
7-Aminoquinazoline-2,4(1H,3H)-dione 95%
7-Aminoquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD13191696
CAS Number: 59674-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR