m-PEG7-alcohol 95%

Supplier: Ambeed
Warning

Synonyms: O-Methyl-heptaethylene glycol, Heptaethylene glycol monomethyl ether

A532586-100MG A532586-1G A532586-250MG A532586-5G A532586-25G
76878-328EA 8.49 USD
76878-328 76878-330 76878-332 76878-334 77613-676
m-PEG7-alcohol 95%
m-PEG7-alcohol

Precaución: for Research Use Only

Formula: C₁₅H₃₂O₈
MW: 340.41 g/mol
MDL Number: MFCD06201006
CAS Number: 4437-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR