2-(Dimethylcarbamoyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

A531404-250MG A531404-100MG
76878-320EA 66 USD
76878-320 77884-318
2-(Dimethylcarbamoyl)phenylboronic acid pinacol ester 98%
2-(Dimethylcarbamoyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BNO₃
MDL Number: MFCD07368236
CAS Number: 956229-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR