Z-Arg(Mtr)-OH CHA 98%

Supplier: Ambeed
Warning

A530713-25G A530713-5G
76878-316EA 62 USD
76878-316 76878-318
Z-Arg(Mtr)-OH CHA 98%
Z-Arg(Mtr)-OH CHA

Precaución: for Research Use Only

Formula: C₂₄H₃₂N₄O₇S·C₆H₁₃N
MDL Number: MFCD00057997
CAS Number: 80745-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR