[(2,6-Dimethylphenyl)amino](oxo)acetic Acid 97%

Supplier: Ambeed
Danger

Synonyms: 2,6-Dimethylanilino(oxo)acetic acid, 2-((2,6-Dimethylphenyl)amino)-2-oxoacetic acid, N-(2,6-Dimethylphenyl)oxamic acid, DMPAO, (2,6-Dimethylanilino)(oxo)acetic acid, [(2,6-Dimethylphenyl)Carbamoyl]formic acid

A529911-10G A529911-1G A529911-250MG A529911-25G A529911-5G
76878-302EA 101 USD
76878-302 76878-304 76878-306 76878-308 76878-310
[(2,6-Dimethylphenyl)amino](oxo)acetic Acid 97%
[(2,6-Dimethylphenyl)amino](oxo)acetic Acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
MDL Number: MFCD10018453
CAS Number: 2903-48-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR