2-(4-Boronophenyl)-2-methylpropanoic acid 95%

Supplier: Ambeed
Warning

A529080-1G A529080-250MG A529080-5G
76878-290EA 201.58 USD
76878-290 76878-292 76878-294
2-(4-Boronophenyl)-2-methylpropanoic acid 95%
2-(4-Boronophenyl)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₄
MW: 208.02 g/mol
MDL Number: MFCD09991614
CAS Number: 1187209-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR