(6-Bromo-2-methylpyridin-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-methylpyridine-3-boronic acid

A528234-1G A528234-250MG A528234-5G
76878-284EA 143.23 USD
76878-284 76878-286 76878-288
(6-Bromo-2-methylpyridin-3-yl)boronic acid 98%
(6-Bromo-2-methylpyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BBrNO₂
MW: 215.84 g/mol
MDL Number: MFCD09037490
CAS Number: 1072944-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR