2-Amino-4-(trifluoromethyl)benzamide 98%

Supplier: Ambeed
Warning

A525819-100MG A525819-250MG
76878-272EA 31 USD
76878-272 76878-274
2-Amino-4-(trifluoromethyl)benzamide 98%
2-Amino-4-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₇F₃N₂O
MDL Number: MFCD06808543
CAS Number: 713-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR