1,4,5,6-Tetrahydro-1-methylpyrrolo[3,4-c]pyrazole hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole hydrochloride

A525695-100MG A525695-1G A525695-250MG
76878-266EA 40.32 USD
76878-266 76878-268 76878-270
1,4,5,6-Tetrahydro-1-methylpyrrolo[3,4-c]pyrazole hydrochloride 97%
1,4,5,6-Tetrahydro-1-methylpyrrolo[3,4-c]pyrazole hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀ClN₃
MW: 159.62 g/mol
MDL Number: MFCD12400786
CAS Number: 1187830-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR