1-(4-(Hydroxymethyl)piperidin-1-yl)ethanone 95%

Supplier: Ambeed
Warning

A525194-1G A525194-25G A525194-5G
76878-254EA 15 USD
76878-254 76878-256 76878-258
1-(4-(Hydroxymethyl)piperidin-1-yl)ethanone 95%
1-(4-(Hydroxymethyl)piperidin-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MDL Number: MFCD11611130
CAS Number: 846057-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR