2,4-Dichloro-5-thiazolecarboxaldehyde 96%

Supplier: Ambeed
Danger

Synonyms: 2,4-Dichlorothiazole-5-carbaldehyde

A522204-1G A522204-5G
76878-238EA 23.34 USD
76878-238 76878-240
2,4-Dichloro-5-thiazolecarboxaldehyde 96%
2,4-Dichloro-5-thiazolecarboxaldehyde

Precaución: for Research Use Only

Formula: C₄HCl₂NOS
MW: 182.03 g/mol
MDL Number: MFCD00793013
CAS Number: 92972-48-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR