6-(Dimethylamino)picolinaldehyde 95%

Supplier: Ambeed
Warning

A520235-1G A520235-250MG
76878-222EA 421 USD
76878-222 77884-300
6-(Dimethylamino)picolinaldehyde 95%
6-(Dimethylamino)picolinaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MDL Number: MFCD11043824
CAS Number: 208110-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR