3-Bromo-2,6-difluorophenol 98%

Supplier: Ambeed
Warning

A518863-1G A518863-250MG A518863-5G
76878-210EA 45 USD
76878-210 76878-212 76878-214
3-Bromo-2,6-difluorophenol 98%
3-Bromo-2,6-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MDL Number: MFCD12910487
CAS Number: 221220-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR