5-Bromo-2-ethoxy-4-methylpyridine 98%

Supplier: Ambeed
Warning

A517616-1G A517616-5G
76878-204EA 75.33 USD
76878-204 76878-206
5-Bromo-2-ethoxy-4-methylpyridine 98%
5-Bromo-2-ethoxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.07 g/mol
MDL Number: MFCD06254609
CAS Number: 610279-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR