2-Benzylmorpholine 95%

Supplier: Ambeed
Warning

A515201-100MG A515201-1G A515201-250MG
76878-196EA 50.93 USD
76878-196 76878-198 76878-200
2-Benzylmorpholine 95%
2-Benzylmorpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MW: 177.243 g/mol
MDL Number: MFCD09055007
CAS Number: 87955-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR