2-Ethylthiazole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A513835-1G A513835-250MG
76878-178EA 322.52 USD
76878-178 76878-180
2-Ethylthiazole-5-carbaldehyde 97%
2-Ethylthiazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₇NOS
MW: 141.19 g/mol
MDL Number: MFCD14588408
CAS Number: 933683-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR