2-Fluoro-5-methoxyphenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-3-hydroxyanisole

A512989-1G A512989-250MG A512989-5G
76878-172EA 111.4 USD
76878-172 76878-174 76878-176
2-Fluoro-5-methoxyphenol 98%
2-Fluoro-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
MW: 142.128 g/mol
MDL Number: MFCD16998752
CAS Number: 117902-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR