Methyl 3-amino-5-chloro-2-methylbenzoate 97%

Supplier: Ambeed
Warning

A512785-1G A512785-250MG A512785-5G
76878-166EA 119.89 USD
76878-166 76878-168 76878-170
Methyl 3-amino-5-chloro-2-methylbenzoate 97%
Methyl 3-amino-5-chloro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.634 g/mol
MDL Number: MFCD23701503
CAS Number: 294190-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR