5-Bromo-1H-pyrrolo[2,3-b]pyridin-4-amine 95%

Supplier: Ambeed
Warning

A512105-1G A512105-100MG
76878-162EA 196.27 USD
76878-162 77673-358
5-Bromo-1H-pyrrolo[2,3-b]pyridin-4-amine 95%
5-Bromo-1H-pyrrolo[2,3-b]pyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD12962871
CAS Number: 1190317-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR