1-Boc-6-bromoindole 96%

Supplier: Ambeed
Danger

A510765-1G A510765-250MG A510765-5G
76878-148EA 67 USD
76878-148 76878-150 76878-152
1-Boc-6-bromoindole 96%
1-Boc-6-bromoindole

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrNO₂
MDL Number: MFCD09954826
CAS Number: 147621-26-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR