Ofloxacin 98+%

Supplier: Ambeed
Warning

Synonyms: 9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid, 9-Fluoro-3,7-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid

A510593-1G A510593-250MG A510593-5G A510593-100G A510593-1MG A510593-25G
76878-142EA 7.43 USD
76878-142 76878-144 76878-146 77673-342 77673-344 77673-346
Ofloxacin 98+%
Ofloxacin

Precaución: for Research Use Only

Formula: C₁₈H₂₀FN₃O₄
MW: 361.37 g/mol
MDL Number: MFCD00226105
CAS Number: 82419-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR