3-(Ethoxycarbonyl)-5-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-(Ethoxycarbonyl)-5-fluorophenyl)boronic acid, 3-Fluoro-5-(ethoxycarbonyl)benzeneboronic acid

A510285-1G A510285-250MG A510285-5G
76878-136EA 80 USD
76878-136 76878-138 76878-140
3-(Ethoxycarbonyl)-5-fluorophenylboronic acid 97%
3-(Ethoxycarbonyl)-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
MDL Number: MFCD07363792
CAS Number: 871329-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR