3-Butoxy-5-formylphenylboronic acid 98+%

Supplier: Ambeed
Danger

Synonyms: (3-Butoxy-5-formylphenyl)boronic acid, 3-butoxy-5-formylphenylboronic acid

A509968-100MG A509968-1G A509968-250MG
76878-130EA 36 USD
76878-130 76878-132 76878-134
3-Butoxy-5-formylphenylboronic acid 98+%
3-Butoxy-5-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₄
MDL Number: MFCD08705271
CAS Number: 1072951-70-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR