1-(3-Bromophenylsulfonyl)pyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-((3-Bromophenyl)sulphonyl)pyrrolidine

A509786-1G A509786-25G A509786-5G
76878-124EA 23.34 USD
76878-124 76878-126 76878-128
1-(3-Bromophenylsulfonyl)pyrrolidine 98%
1-(3-Bromophenylsulphonyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂S
MW: 290.1768 g/mol
MDL Number: MFCD07363829
CAS Number: 214210-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR