8-Methyl-2-quinolinol 97%

Supplier: Ambeed
Warning

Synonyms: 8-Methylquinolin-2(1H)-one, 8-Methyl-2(1H)-quinolinone

A508722-1G A508722-5G
76878-114EA 33 USD
76878-114 76878-116
8-Methyl-2-quinolinol 97%
8-Methyl-2-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD13195790
CAS Number: 4053-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR