5-Ethynyl-2-methoxypyridine 95%

Supplier: Ambeed
Warning

A508182-100MG A508182-1G A508182-250MG A508182-25G A508182-5G
76878-104EA 25 USD
76878-104 76878-106 76878-108 76878-110 76878-112
5-Ethynyl-2-methoxypyridine 95%
5-Ethynyl-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₈H₇NO
MDL Number: MFCD10698135
CAS Number: 663955-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR