3-Bromo-2-(methylthio)-5-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Danger

A507615-1G A507615-250MG A507615-5G
76878-098EA 82.76 USD
76878-098 76878-100 76878-102
3-Bromo-2-(methylthio)-5-(trifluoromethyl)pyridine 97%
3-Bromo-2-(methylthio)-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NS
MW: 272.0855 g/mol
MDL Number: MFCD16295130
CAS Number: 1226808-64-5
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR