2,4-Difluoro-3-(trifluoromethyl)bromobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2,6-difluorobenzotrifluoride, 1-Bromo-2,4-difluoro-3-(trifluoromethyl)benzene

A507151-100MG A507151-1G A507151-250MG
76878-090EA 24.41 USD
76878-090 76878-092 76878-094
2,4-Difluoro-3-(trifluoromethyl)bromobenzene 98%
2,4-Difluoro-3-(trifluoromethyl)bromobenzene

Precaución: for Research Use Only

Formula: C₇H₂BrF₅
MW: 260.99 g/mol
MDL Number: MFCD18379808
CAS Number: 1263377-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR