Cilostazol 98%

Supplier: Ambeed
Danger

A506538-100MG A506538-1G A506538-250MG A506538-50MG A506538-5G A506538-25G
76878-078EA 6 USD
76878-078 76878-080 76878-082 76878-084 77673-326 77884-274
Cilostazol 98%
Cilostazol

Precaución: for Research Use Only

Formula: C₂₀H₂₇N₅O₂
MDL Number: MFCD00866780
CAS Number: 73963-72-1
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR