N1-(2-(4-(2-Aminoethyl)piperazin-1-yl)ethyl)ethane-1,2-diamine 95%

Supplier: Ambeed
Danger

A505336-1G A505336-5G A505336-100MG A505336-250MG
76878-074EA 315.09 USD
76878-074 76878-076 77568-986 77613-596
N1-(2-(4-(2-Aminoethyl)piperazin-1-yl)ethyl)ethane-1,2-diamine 95%
N1-(2-(4-(2-Aminoethyl)piperazin-1-yl)ethyl)ethane-1,2-diamine

Precaución: For research use only.

Formula: C₁₀H₂₅N₅
MW: 215.33 g/mol
MDL Number: MFCD01941014
CAS Number: 31295-54-2
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR