3-Chloro-o-toluidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-chlorotoluene, 3-Chloro-2-methylaniline

A503859-100G A503859-10G A503859-25G A503859-500G
76878-060EA 20.16 USD
76878-060 76878-062 76878-064 76878-066
3-Chloro-o-toluidine 98%
3-Chloro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.6 g/mol
MDL Number: MFCD00007766
CAS Number: 87-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR