1-Boc-7-methoxyindole 96%

Supplier: Ambeed
Warning

A498850-1G A498850-250MG A498850-5G
76878-032EA 34 USD
76878-032 76878-034 76878-036
1-Boc-7-methoxyindole 96%
1-Boc-7-methoxyindole

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MDL Number: MFCD15143582
CAS Number: 1215205-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR