3-Chloro-2-cyanobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-2-cyanophenyl)boronic acid

A496015-1G A496015-250MG A496015-5G
76878-014EA 200.52 USD
76878-014 76878-016 76878-018
3-Chloro-2-cyanobenzeneboronic acid 98%
3-Chloro-2-cyanobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClNO₂
MW: 181.382 g/mol
MDL Number: MFCD10696670
CAS Number: 1217500-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR