3-Bromo-6-(trifluoromethyl)pyridin-2-amine 98%

Supplier: Ambeed
Warning

A494568-100MG A494568-1G A494568-250MG A494568-25G A494568-5G
76877-996EA 23 USD
76877-996 76877-998 76878-000 76878-002 76878-004
3-Bromo-6-(trifluoromethyl)pyridin-2-amine 98%
3-Bromo-6-(trifluoromethyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₄BrF₃N₂
MW: 241.00 g/mol
MDL Number: MFCD13185328
CAS Number: 1214361-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR