Naphthalene-2,7-diamine 95%

Supplier: Ambeed
Warning

A493385-100MG A493385-1G A493385-250MG A493385-50MG
76877-982EA 33 USD
76877-982 76877-984 76877-986 76877-988
Naphthalene-2,7-diamine 95%
Naphthalene-2,7-diamine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MDL Number: MFCD00021651
CAS Number: 613-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR